Benzoin ethyl ether

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Benzoin Ethyl EtherO-Ethylbenzoin

11280
100503-684EA 76.13 USD
100503-684
Benzoin ethyl ether
Benzoin ethyl ether
Formula: C₁₆H₁₆O₂
MW: 240.3 g/mol
MDL Number: MFCD00009242
CAS Number: 574-09-4
https://us.vwr.com/stibo/web/std.lang.all/47/85/28984785.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR