Basic fuchsin, certified CI 42510, CN DcFb-26
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
Basic-fuchsin
, Pararosaniline
, Fuchsin basic
, Basic fuchsin hydrochloride
, Diamondfuchsin
, Basic Red 9
, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride
, Basic fuchsin
, CI 42510
, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride
, Magenta O
, Rosaniline hydrochloride
Can be used for gram positive/negative bacteria and as a pituitary stain.
Can also be used in the selective staining of nervous tissue for light microscopy following preparation for electron microscopy (Berkowitz et al., J. Histochem. Cytochem. 16:808, 1968.
Can be used as a simple rapid polychrome stain for epoxy-embedded tissue (Sato, T., and Shamoto, M. Stain Technol. 48:223, 1973).
Certifications: Certified by the Biological Stain Commission.
Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.
Formula:
C₂₀H₂₀ClN₃ MW: 337.85 g/mol Melting Pt: 205 °C (401 °F) |
MDL Number:
MFCD00012569 CAS Number: 632-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...