Azur II eosinate (C.I. 52010/52015/45380)

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Azure II eosinDipotassium 7-(dimethylamino)-N,N-dimethylphenothiazin-3-iminium N,N-dimethyl-7-(methylamino)phenothiazin-3-iminium eosin YS(2-) dichloride

11220
100503-674EA 95.71 USD
100503-674
Azur II eosinate (C.I. 52010/52015/45380)
Azur II eosinate

Azur B/Methylene Blue/Eosin 1:1:1.

Formula: C₅₁H₄₀Br₄Cl₂N₆Na₂O₅S₂
MW: 1317.534 g/mol
MDL Number: MFCD00084753
CAS Number: 53092-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR