Azure A, certified used in nuclear staining C.I. 52005

Supplier: Electron Microscopy Sciences
Warning

Synonyms: N,N-DimethylthionineMethylene Azur A

11240
100503-676EA 118.17 USD
100503-676
Azure A, certified used in nuclear staining C.I. 52005
Azure A

Can be used in nuclear staining; A polychromatic blood stain.


Solubility : Water-soluble; Alcohol-soluble


Certifications: Certified by the Biological Stain Commission.


Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.

Formula: C₁₄H₁₄ClN₃S
MW: 291.797 g/mol
MDL Number: MFCD00012112
CAS Number: 531-53-3
https://us.vwr.com/stibo/web/std.lang.all/48/56/28984856.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR