Aniline blue, certified (C.I. 42755, C.N. DcK-11)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Acid Blue 22Marine Blue VDisodium hydrogen aminomethyl[[4-[(sulphonatophenyl)amino]phenyl][4-[(sulphonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulphonateChina BlueCotton BlueCI 42755Blackley BlueCN DcK-11Water Blue

10840
100503-664EA 48.3 USD
100503-664
Aniline blue, certified (C.I. 42755, C.N. DcK-11)
Aniline blue

Used in connective tissue staining procedures to stain collagen. Used in differential staining (Snodgress el al., Lab Invest. 26:329, 1972).


Can be used with biebrich scarlet for staining collagen, reticulum, muscle, plasma, and nuclei. All connective tissues.


For use in staining cytoplasm, cellulose and determining hydrolysis of fats. Can also be used as a contrast stain for carmine and as a counterstain with red nuclear dye.


Solubility : 50% Water; 0.0% Alcohol; 4.5% Cellosolve; 7.8% Glycol; 0.0% Xylene


Certifications: Certified by the Biological Stain Commission.


Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.

Formula: C₃₂H₂₅N₃Na₂O₉S₃
MW: 737.74 g/mol
MDL Number: MFCD00036130
CAS Number: 28631-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR