2-Dimethylaminoethyl methacrylate
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
DMA
, N,N-Dimethylaminoethyl methacrylate
13350
100503-828EA
28.15
USD
100503-828
2-Dimethylaminoethyl methacrylate
2-Dimethylaminoethyl methacrylate
A water soluble monomer.
Formula:
C₈H₁₅NO₂ MW: 157.21 g/mol Boiling Pt: 182…190 °C (360…374 °F) Melting Pt: –30.00 °C Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008589 CAS Number: 2867-47-2 UN: 2522 ADR: 6.1,II |
Specification Test Results
Specific Gravity | 0.933 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...