Butyl glycidyl ether

Supplier: Electron Microscopy Sciences
Warning

Synonyms: BGEButoxymethyl oxiraneAraldite RD-1

12000
100504-924EA 48.64 USD
100504-924
Butyl glycidyl ether
Butyl glycidyl ether
Formula: C₇H₁₄O₂
Density: 0.92 g/cm³ (7.677 lbs/gal.)
Flash Pt: 56 °C (133 °F)
MDL Number: MFCD00005145
CAS Number: 2426-08-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Specific gravity 0.912

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR