Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hemoglobin A1c Reagent Set, Pointe Scientific

Supplier: POINTE SCIENTIFIC, INC.
H7546-40 H7546-120 OH546-40
For the quantitative determination of Hemoglobin A1c (HbA1c) in human blood

This method utilizes the interaction of antigen and antibody to directly determine the HbA1c in whole blood. Total hemoglobin and HbA1c have the same unspecific absorption rate to latex particles. When mouse antihuman HbA1c monoclonal antibody is added (R2), latex-HbA1c-mouse anti human HbA1c antibody complex is formed. Agglutination is formed when goat anti-mouse IgG polyclonal antibody interacts with the monoclonal antibody. The amount of agglutination is proportional to the amount of HbA1c absorbed on to the surface of latex particles. The amount of agglutination is measured as absorbance. The HbA1c value is obtained from a calibration curve.

The Olympus Hemoglobin A1c reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers.

The determination of HbA1c is most commonly performed for the evaluation of glycemic control in diabetes mellitus. HbA1c values provide an indication of glucose levels over the preceding 4-8 weeks. A higher HbA1c value indicates poorer glycemic control. For in vitro diagnostic use only.

Certifications: CE Compliant, Health Canada (For OH546-40 Only)

Ordering information: Hemoglobin A1c calibrator set, control set, and Hemolysis Reagent are required but not provided (Provided in the Reagent Kit only).



 

ORDER

Expand All / Collapse All
Description Format Includes Storage Temperature Supplier No. VWR Catalog Number Unit Price Quantity
Hemoglobin A1c Reagent Kit Liquid 90 mL R1 Reagent and 30 mL R2 Reagent 2 to 8°C H7546-120 10022-854 Each (120ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Hemoglobin A1c Reagent Kit Liquid 30 mL R1 Reagent, 10 mL R2 Reagent and 125 mL Lyse Reagent 2 to 8°C H7546-40 10022-852 Each (40ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Olympus Hemoglobin A1c Reagent Liquid 30 mL R1 Reagent and 10 mL R2 Reagent 2 to 8°C OH546-40 10023-488 Each (40ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Hemoglobin A1c Lyse Reagent, 500 mL, Liquid, Store at 15 to 30 °C H7541-LYS 10022-840 Each (500ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty