Would you like to log in again?
Anatomical Pathology Tissue and Embedding Cassette, Thermo Scientific
Supplier: Thermo Scientific
B1003500OR B1000732YW B1003500GR B1000732OR B1000732WH B1003500WH B1003500YW B1000732LI B1000732PK 58341 58343 58342 B1000732BL B1003500BL B1003500PE 58406 B1000732GR B1000732GY B1003500AQ B1000732AQ 58402 58401 B1003500GY B1003500PK B1000732PE B1003500LI B1000732TN 58408 58407 B1003500TN
Hold and identify tissue samples in processing, embedding and sectioning procedures with Thermo Scientific™ Shandon™ Embedding Cassettes in Tube Packs.
  • Can be used on all Thermo Scientific automatic cassette labelers
  • Fit securely in microtome chuck adapters
  • Designed to accept metal lids
  • Made of acetal polymer with a pore style of round holes
  • Embedding cassettes supplied in 10 tubes of 75 cassettes each
  • Tissue cassettes designed to accept metal lids
  • Designed with round hole pores
  • Supplied in boxes of 500 cassettes
Thermo Scientific® Shandon and Richard-Allan Scientific Embedding cassettes are made from chemical resistant polymers which withstand the normal histology process including microwave tissue processing. All cassettes fit standard microtome block holders. Cassette pore styles include slotted, small square, mesh, small holes (0.36mm diameter), and round. Many are compatible with automatic labelers. Writing surfaces are a flat forty-five degree angle, and lids are either attached, included but not attached, or ordered separately. Thermo Scientific® Shandon and Richard-Allan Scientific Tissue Cassettes are designed with round hole pores and to accept metal lids.
ORDER
Expand All / Collapse All
Description Color Supplier No. VWR Catalog Number Unit Price Quantity
Embedding Cassette Aqua B1003500AQ 89400-130 Case of 500 Retrieving Restricted
Embedding Cassette Aqua B1000732AQ 89399-996 Case of 750 Retrieving Restricted
Embedding Cassette Blue B1003500BL 89400-132 Case of 500 Retrieving Restricted
Embedding Cassette Blue B1000732BL 89399-998 Case of 750 Retrieving Restricted
Embedding Cassette Green B1003500GR 89400-134 Case of 500 Retrieving Restricted
Embedding Cassette Green B1000732GR 89400-000 Case of 750 Retrieving Restricted
Embedding Cassette Grey B1003500GY 89400-136 Case of 500 Retrieving Restricted
Embedding Cassette Grey B1000732GY 89400-002 Case of 750 Retrieving Restricted
Embedding Cassette Lilac B1003500LI 89400-138 Case of 500 Retrieving Restricted
Embedding Cassette Lilac B1000732LI 89400-004 Case of 750 Retrieving Restricted
Embedding Cassette Orange B1003500OR 89400-140 Case of 500 Retrieving Restricted
Embedding Cassette Orange B1000732OR 89400-006 Case of 750 Retrieving Restricted
Embedding Cassette Peach B1003500PE 89400-142 Case of 500 Retrieving Restricted
Embedding Cassette Peach B1000732PE 89400-008 Case of 750 Retrieving Restricted
Embedding Cassette Pink B1003500PK 89400-144 Case of 500 Retrieving Restricted
Embedding Cassette Pink B1000732PK 89400-010 Case of 750 Retrieving Restricted
Embedding Cassette Tan B1003500TN 89400-146 Case of 500 Retrieving Restricted
Embedding Cassette Tan B1000732TN 89400-012 Case of 750 Retrieving Restricted
Embedding Cassette White B1003500WH 89400-148 Case of 500 Retrieving Restricted
Embedding Cassette White B1000732WH 89400-014 Case of 750 Retrieving Restricted
Embedding Cassette Yellow B1003500YW 89400-150 Case of 500 Retrieving Restricted
Embedding Cassette Yellow B1000732YW 89400-016 Case of 750 Retrieving Restricted
Tissue Cassette IV Aqua 58341 72550-022 Case of 1,000 Retrieving Restricted
Tissue Cassette IV Blue 58402 72550-014 Case of 1,000 Retrieving Restricted
Tissue Cassette IV Gold 58342 72550-024 Case of 1,000 Retrieving Restricted
Tissue Cassette IV Grey 58408 72550-034 Case of 1,000 Retrieving Restricted
Tissue Cassette IV Lilac 58343 72550-028 Case of 1,000 Retrieving Restricted
Tissue Cassette IV Peach 58407 72550-030 Case of 1,000 Retrieving Restricted
Tissue Cassette IV Tan 58406 72550-032 Case of 1,000 Retrieving Restricted
Tissue Cassette IV White 58401 72550-012 Case of 1,000 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty