Merit™ Schroeder Tenaculum Forceps, Physician Grade, Sklar
Supplier: Sklar
98-386
10185-326EA 41.74 USD
10185-326
Merit™ Schroeder Tenaculum Forceps, Physician Grade, Sklar
Forceps
Merit™ Schroeder-Braun Tenaculum Forceps are a ratcheted, finger ring instrument used in OB/GYN procedures involving manipulation of the cervix or uterus, D&C, and vaginal or abdominal hysterectomy
They are used to grasp, hold, and move vessels and arteries during surgical procedures.