Water, HiPerSolv CHROMANORM® for LC-MS, VWR Chemicals BDH®
Synonyms:
Dihydrogen oxide
BDHVBDH83645.400
BDH83645.400CS
304.3
USD
BDH83645.400
Water, HiPerSolv CHROMANORM® for LC-MS, VWR Chemicals BDH®
Water
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 12,10175 |
Specification Test Results
Colouration | Max. 5 APHA |
Conductivity (25°C) (at filling) | Max. 1 µS/cm |
Evaporation residue | Max. 1 ppm |
Al (Aluminium) | Max. 50 ppb |
Ca (Calcium) | Max. 50 ppb |
Fe (Iron) | Max. 50 ppb |
K (Potassium) | Max. 50 ppb |
Mg (Magnesium) | Max. 50 ppb |
Na (Sodium) | Max. 100 ppb |
Fluorescence (as quinine) (254/450 nm) | Max. 1 ppb |
Fluorescence (as quinine) (365/450 nm) | Max. 0,5 ppb |
Gradient grade (210 nm) | Max. 5 mAU |
Gradient grade (220 nm) | Max. 2 mAU |
Gradient grade (254 nm) | Max. 1 mAU |
Max. 2 ppb | Max. 2 ppb |
Mode: ESI 200 µl neg | APCI 200 µl | Max. 20 ppb |
Colony count (Ph.Eur.) | Max. 100 CFU/ml |
TOC (at filling) | Max. 30 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...