Anti-NME2 Mouse Monoclonal Antibody [clone: 1F2]

Supplier: Abnova
H00004831-M08A
10625-414EA 335 USD
10625-414
Anti-NME2 Mouse Monoclonal Antibody [clone: 1F2]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant NME2.

Type: Primary
Antigen: NME2
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR