Anti-NEU1 Mouse Monoclonal Antibody [clone: 3F9]

Supplier: Abnova
H00004758-M01
10594-700EA 335 USD
10594-700
Anti-NEU1 Mouse Monoclonal Antibody [clone: 3F9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant NEU1.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: NEU1
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR