Anti-MYO1E Mouse Monoclonal Antibody [clone: 7A5]

Supplier: Abnova
H00004643-M02
10594-206EA 335 USD
10594-206
Anti-MYO1E Mouse Monoclonal Antibody [clone: 7A5]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant MYO1E.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: MYO1E
Clonality: Monoclonal
Clone: 7A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR