Anti-MATK Mouse Monoclonal Antibody [clone: 2C5]

Supplier: Abnova
H00004145-M05A
10623-972EA 335 USD
10623-972
Anti-MATK Mouse Monoclonal Antibody [clone: 2C5]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant MATK.

Type: Primary
Antigen: MATK
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR