Anti-LMNB1 Mouse Monoclonal Antibody [clone: 4B10]

Supplier: Abnova
H00004001-M01A
10624-560EA 335 USD
10624-560
Anti-LMNB1 Mouse Monoclonal Antibody [clone: 4B10]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant LMNB1.

Type: Primary
Antigen: LMNB1
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR