Anti-KCNF1 Mouse Monoclonal Antibody [clone: S2]

Supplier: Abnova
H00003754-M01A
10624-778EA 335 USD
10624-778
Anti-KCNF1 Mouse Monoclonal Antibody [clone: S2]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant KCNF1.

Type: Primary
Antigen: KCNF1
Clonality: Monoclonal
Clone: S2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR