Anti-KCNE1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00003753-D01
10590-450EA 376 USD
10590-450
Anti-KCNE1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human KCNE1 protein.

Recommended Dilutons: Western Blot (1:1000)

Type: Primary
Antigen: KCNE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR