Anti-ING2 Mouse Polyclonal Antibody

Supplier: Abnova
H00003622-B01
10561-802EA 335 USD
10561-802
Anti-ING2 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human ING2 protein.

Recommended Dilutons: Western Blot (1:500-1:1000)

Type: Primary
Antigen: ING2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR