Anti-ING1 Mouse Monoclonal Antibody [clone: 2F9]

Supplier: Abnova
H00003621-M05A
10624-532EA 335 USD
10624-532
Anti-ING1 Mouse Monoclonal Antibody [clone: 2F9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant ING1.

Type: Primary
Antigen: ING1
Clonality: Monoclonal
Clone: 2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR