Anti-IL1RN Rabbit Polyclonal Antibody

Supplier: Abnova
H00003557-D01
10590-258EA 376 USD
10590-258
Anti-IL1RN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human IL1RN protein.

Recommended Dilutons: Western Blot (1:1000)

Type: Primary
Antigen: IL1RN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR