Anti-IGLC2 Mouse Polyclonal Antibody

Supplier: Abnova
H00003538-B01P
10561-520EA 335 USD
10561-520
Anti-IGLC2 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human IGLC2 protein.

Recommended Dilutons: Western Blot (1 ug/mL)

Type: Primary
Antigen: IGLC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR