Anti-HOXB6 Mouse Monoclonal Antibody [clone: 8000]

Supplier: Abnova
H00003216-M01
10589-210EA 335 USD
10589-210
Anti-HOXB6 Mouse Monoclonal Antibody [clone: 8000]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant HOXB6.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: HOXB6
Clonality: Monoclonal
Clone: 8000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR