Anti-HARS Mouse Monoclonal Antibody [clone: 4D4]

Supplier: Abnova
H00003035-M03A
10624-406EA 335 USD
10624-406
Anti-HARS Mouse Monoclonal Antibody [clone: 4D4]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant HARS.

Type: Primary
Antigen: HARS
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR