Anti-BRF1 Mouse Monoclonal Antibody [clone: 2E9]

Supplier: Abnova
H00002972-M01A
10623-858EA 335 USD
10623-858
Anti-BRF1 Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant BRF1.

Type: Primary
Antigen: BRF1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR