Anti-GRP Mouse Monoclonal Antibody [clone: 3A11]

Supplier: Abnova
H00002922-M03
10587-970EA 335 USD
10587-970
Anti-GRP Mouse Monoclonal Antibody [clone: 3A11]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant GRP.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: GRP
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR