Anti-GFAP Mouse Monoclonal Antibody [clone: 8H3]

Supplier: Abnova
H00002670-M06
10586-414EA 335 USD
10586-414
Anti-GFAP Mouse Monoclonal Antibody [clone: 8H3]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant GFAP.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 8H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR