Anti-GATA4 Mouse Monoclonal Antibody [clone: 6C6]

Supplier: Abnova
H00002626-M01
10587-124EA 335 USD
10587-124
Anti-GATA4 Mouse Monoclonal Antibody [clone: 6C6]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant GATA4.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: GATA4
Clonality: Monoclonal
Clone: 6C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR