Anti-FLI1 Mouse Monoclonal Antibody [clone: 1H4]

Supplier: Abnova
H00002313-M07
10585-388EA 335 USD
10585-388
Anti-FLI1 Mouse Monoclonal Antibody [clone: 1H4]
Antibodies
Mouse monoclonal antibody raised against a full length recombinant FLI1.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: FLI1
Clonality: Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR