Anti-FOXO3 Mouse Monoclonal Antibody [clone: 1E11]

Supplier: Abnova
H00002309-M13A
10624-340EA 335 USD
10624-340
Anti-FOXO3 Mouse Monoclonal Antibody [clone: 1E11]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant FOXO3.

Type: Primary
Antigen: FOXO3
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR