Anti-ETV6 Mouse Polyclonal Antibody

Supplier: Abnova
H00002120-A01
10556-100EA 243 USD
10556-100
Anti-ETV6 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length recombinant ETV6.

Recommended Dilutons: Western Blot (1:500-1:1000)

Type: Primary
Antigen: ETV6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR