Anti-ETV1 Mouse Monoclonal Antibody [clone: 2A8]

Supplier: Abnova
H00002115-M01
10585-284EA 335 USD
10585-284
Anti-ETV1 Mouse Monoclonal Antibody [clone: 2A8]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant ETV1.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: ETV1
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR