Anti-SERPINB1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00001992-D01P
10600-804EA 376 USD
10600-804
Anti-SERPINB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human SERPINB1 protein.

Recommended Dilutons: Western Blot (1 ug/mL)

Type: Primary
Antigen: SERPINB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR