Anti-DDX5 Mouse Polyclonal Antibody

Supplier: Abnova
H00001655-B01
10554-876EA 335 USD
10554-876
Anti-DDX5 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human DDX5 protein.

Recommended Dilutons: Western Blot (1:500-1:1000)

Type: Primary
Antigen: DDX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR