Anti-COL9A3 Mouse Monoclonal Antibody [clone: 2B5]

Supplier: Abnova
H00001299-M02A
10624-102EA 335 USD
10624-102
Anti-COL9A3 Mouse Monoclonal Antibody [clone: 2B5]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant COL9A3.

Type: Primary
Antigen: COL9A3
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR