Anti-CDX4 Mouse Monoclonal Antibody [clone: 2H7]

Supplier: Abnova
H00001046-M11
10581-896EA 335 USD
10581-896
Anti-CDX4 Mouse Monoclonal Antibody [clone: 2H7]
Antibodies
Mouse monoclonal antibody raised against a full length recombinant CDX4.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: CDX4
Clonality: Monoclonal
Clone: 2H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR