Anti-CD59 Mouse Polyclonal Antibody

Supplier: Abnova
H00000966-B04
10560-194EA 335 USD
10560-194
Anti-CD59 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human CD59 protein.

Recommended Dilutons: Western Blot (1:500-1:1000)

Type: Primary
Antigen: CD59
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR