Anti-CD58 Mouse Polyclonal Antibody

Supplier: Abnova
H00000965-B01
10560-192EA 335 USD
10560-192
Anti-CD58 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human CD58 protein.

Recommended Dilutons: Western Blot (1:500-1:1000)

Type: Primary
Antigen: CD58
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR