Anti-CD44 Mouse Monoclonal Antibody [clone: 4H2]

Supplier: Abnova
H00000960-M16
10581-358EA 335 USD
10581-358
Anti-CD44 Mouse Monoclonal Antibody [clone: 4H2]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant CD44.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR