Anti-CD40 Mouse Monoclonal Antibody [clone: 3G10]

Supplier: Abnova
H00000958-M20
10594-362EA 335 USD
10594-362
Anti-CD40 Mouse Monoclonal Antibody [clone: 3G10]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant CD40.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR