Anti-CASP4 Mouse Monoclonal Antibody [clone: 7G7]

Supplier: Abnova
H00000837-M02
10578-678EA 335 USD
10578-678
Anti-CASP4 Mouse Monoclonal Antibody [clone: 7G7]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant CASP4.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: CASP4
Clonality: Monoclonal
Clone: 7G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR