Anti-CAMK2B Mouse Polyclonal Antibody

Supplier: Abnova
H00000816-B01
10552-388EA 335 USD
10552-388
Anti-CAMK2B Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human CAMK2B protein.

Recommended Dilutons: Western Blot (1:500-1:1000)

Type: Primary
Antigen: CAMK2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR