Anti-CACNB3 Rabbit Polyclonal Antibody

Supplier: Abnova
H00000784-D01P
10578-470EA 376 USD
10578-470
Anti-CACNB3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human CACNB3 protein.

Recommended Dilutons: Western Blot (1 ug/mL)

Type: Primary
Antigen: CACNB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR