Anti-BUB1B Mouse Monoclonal Antibody [clone: 2G9]

Supplier: Abnova
H00000701-M01A
10623-864EA 335 USD
10623-864
Anti-BUB1B Mouse Monoclonal Antibody [clone: 2G9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant BUB1B.

Type: Primary
Antigen: BUB1B
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR