Anti-BMP5 Mouse Monoclonal Antibody [clone: 4A3]

Supplier: Abnova
H00000653-M30C
10623-754EA 335 USD
10623-754
Anti-BMP5 Mouse Monoclonal Antibody [clone: 4A3]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant BMP5.

Recommended Dilutons: Western Blot (1:500-1:1000)

Type: Primary
Antigen: BMP5
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR