Anti-BMP2 Mouse Monoclonal Antibody [clone: 1A11]

Supplier: Abnova
H00000650-M02
10577-958EA 325 USD
10577-958
Anti-BMP2 Mouse Monoclonal Antibody [clone: 1A11]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant BMP2.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: BMP2
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR