Anti-BLM Mouse Monoclonal Antibody [clone: 1E4]

Supplier: Abnova
H00000641-M01
10577-942EA 335 USD
10577-942
Anti-BLM Mouse Monoclonal Antibody [clone: 1E4]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant BLM.

Recommended Dilutons: Western Blot (1-5 ug/mL)

Type: Primary
Antigen: BLM
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR