Anti-APOA1 Mouse Monoclonal Antibody [clone: 3A11-1A9]

Supplier: Abnova
H00000335-M01A
10616-676EA 335 USD
10616-676
Anti-APOA1 Mouse Monoclonal Antibody [clone: 3A11-1A9]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant APOA1.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: 3A11-1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR