Anti-AMELX Mouse Monoclonal Antibody [clone: 5B2]

Supplier: Abnova
H00000265-M02A
10615-582EA 335 USD
10615-582
Anti-AMELX Mouse Monoclonal Antibody [clone: 5B2]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant AMELX.

Type: Primary
Antigen: AMELX
Clonality: Monoclonal
Clone: 5B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR