Anti-AKT1 Mouse Monoclonal Antibody [clone: 4C3]

Supplier: Abnova
H00000207-M01A
10612-358EA 335 USD
10612-358
Anti-AKT1 Mouse Monoclonal Antibody [clone: 4C3]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant AKT1.

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR